Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FIG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568477
Description
FIG4 Polyclonal Antibody for Western Blot
The protein encoded by this gene belongs to the SAC domain-containing protein gene family. The SAC domain, approximately 400 amino acids in length and consisting of seven conserved motifs, has been shown to possess phosphoinositide phosphatase activity. The yeast homolog, Sac1p, is involved in the regulation of various phosphoinositides, and affects diverse cellular functions such as actin cytoskeleton organization, Golgi function, and maintenance of vacuole morphology. Membrane-bound phosphoinositides function as signaling molecules and play a key role in vesicle trafficking in eukaryotic cells. Mutations in this gene have been associated with Charcot-Marie-Tooth disease, type 4J.
Specifications
FIG4 | |
Polyclonal | |
Unconjugated | |
FIG4 | |
A530089I17Rik; AI326867; ALS11; BTOP; CMT4J; dJ249I4.1; FIG4; FIG4 homolog (S. cerevisiae); FIG4 homolog, SAC domain containing lipid phosphatase; FIG4 homolog, SAC1 domain containing lipid phosphatase; FIG4 homolog, SAC1 lipid phosphatase domain containing; FIG4 phosphoinositide 5-phosphatase; KIAA0274; Phosphatidylinositol 3,5-bisphosphate 5-phosphatase; polyphosphoinositide phosphatase; RGD1311375; RP1-249I4.1; Sac domain-containing inositol phosphatase 3; SAC domain-containing protein 3; Sac3; YVS | |
Rabbit | |
affinity chromatography | |
RUO | |
103199, 9896 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q91WF7, Q92562 | |
FIG4 | |
Synthesized peptide derived from human FIG4 [aa 341-390]. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction