Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120353 Supplier Invitrogen™ Supplier No. PA5120353
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, HT-29, Jukat, HeLa, Mouse liver, Mouse spleen. The target is usually found in the following locations: Endosome membrane. Immunogen sequence: WELPTDFYLH HKNTMRLLPT RRSYTYWWTP EVIKHLPLPY DEVICAVNLK KLIVKKFHKY EEEIDIHNEF FRPYELSSFD DTFCLAMTSS ARDFMPKTVG IDPSPFTVRK PDETGKSVLG NKSNREEAVL QRKTAASAPP PPSEEAVSSS SEDDSGTDRE EEGSVSQRST PVKMTDAGDS AKVTENVVQP MKELYGINLS DGLSEEDFSI YSRFVQLGQS QHKQDKNSQQ PCSRCSDGVI KLTPISAFSQ DNIYEVQPPR VDRKSTEIFQ AHIQASQGIM QPLGKEDSSM YREYIRNRYL.

The protein encoded by this gene belongs to the SAC domain-containing protein gene family. The SAC domain, approximately 400 amino acids in length and consisting of seven conserved motifs, has been shown to possess phosphoinositide phosphatase activity. The yeast homolog, Sac1p, is involved in the regulation of various phosphoinositides, and affects diverse cellular functions such as actin cytoskeleton organization, Golgi function, and maintenance of vacuole morphology. Membrane-bound phosphoinositides function as signaling molecules and play a key role in vesicle trafficking in eukaryotic cells. Mutations in this gene have been associated with Charcot-Marie-Tooth disease, type 4J.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIG4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FIG4
Gene Accession No. Q91WF7, Q92562
Gene Alias A530089I17Rik; AI326867; ALS11; BTOP; CMT4J; dJ249I4.1; FIG4; FIG4 homolog (S. cerevisiae); FIG4 homolog, SAC domain containing lipid phosphatase; FIG4 homolog, SAC1 domain containing lipid phosphatase; FIG4 homolog, SAC1 lipid phosphatase domain containing; FIG4 phosphoinositide 5-phosphatase; KIAA0274; Phosphatidylinositol 3,5-bisphosphate 5-phosphatase; polyphosphoinositide phosphatase; RGD1311375; RP1-249I4.1; Sac domain-containing inositol phosphatase 3; SAC domain-containing protein 3; Sac3; YVS
Gene Symbols FIG4
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103199, 9896
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.