Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Figla Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586034 Supplier Invitrogen™ Supplier No. PA586034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Figla Polyclonal Antibody for ICC/IF

Germline specific transcription factor implicated in postnatal oocyte-specific gene expression. Plays a key regulatory role in the expression of multiple oocyte-specific genes, including those that initiate folliculogenesis and those that encode the zona pellucida (ZP1, ZP2 and ZP3) required for fertilization and early embryonic survival. Essential for oocytes to survive and form primordial follicles. The persistence of FIGLA in adult females suggests that it may regulate additional pathways that are essential for normal ovarian development. Binds to the E-box (5'-CANNTG-3') of the ZPs (ZP1, ZP2, ZP3) promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Figla
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene FIGLA
Gene Accession No. O55208, Q6QHK4
Gene Alias bHLHc8; Class C basic helix-loop-helix protein 8; factor in the germline alpha; FIG alpha; FIGalpha; Figla; folliculogenesis specific basic helix-loop-helix; folliculogenesis specific bHLH transcription factor; folliculogenesis-specific basic helix-loop-helix protein; POF6; RGD1564754; transcription factor FIGa
Gene Symbols FIGLA
Host Species Rabbit
Immunogen Synthetic peptide within Human Figla aa 21-70.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26910, 344018
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.