Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIGNL1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002355AF750
Description
FIGNL1 Polyclonal antibody specifically detects FIGNL1 in Human, Mouse samples. It is validated for Western BlotSpecifications
FIGNL1 | |
Polyclonal | |
Western Blot | |
EC 3.6.4, EC 3.6.4.-, fidgetin-like 1, fidgetin-like protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Fidgetin-Like 1using the numbering given in entry NP_071399.2 (GeneID 63979). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
63979 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction