Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIGNL1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326083H
Description
FIGNL1 Polyclonal antibody specifically detects FIGNL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FIGNL1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.6.4, EC 3.6.4.-, fidgetin-like 1, fidgetin-like protein 1 | |
Produced in rabbits immunized with E. coli-derived Human FIGNL1 fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
63979 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction