Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIGNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563140 Supplier Invitrogen™ Supplier No. PA563140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLYIPLPEAS ARKQIVINLM SKEQCCLSEE EIEQIVQQSD AFSGADMTQL CREASLGPIR SLQTADIATI TPDQVRPIAY IDFENAFRTV RPSVSPK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.

The mammalian Phospholipase Cg (PLCg) family has two closely related proteins, PLCg1 and PLCg2. The PLCg isozymes have the core structure domains and a unique array of domains containing an additional PH domain, two SH2 domains and one SH3 domain. In response to extracellular stimuli, such as hormones and growth factors, receptor tyrosine kinases (RTKs) phosphorylate and activate PLCg. Activated PLCg catalyzes hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) to produce the metabolic second messenger molecules inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIGNL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FIGNL1
Gene Accession No. Q6PIW4
Gene Alias fidgetin like 1; fidgetin-like 1; fidgetin-like protein 1; FIGNL1; S30
Gene Symbols FIGNL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FIGNL1. Recombinant protein control fragment (Product #RP-92719).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.