Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIGNL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 176041AP150UL
Description
The mammalian Phospholipase Cg (PLCg) family has two closely related proteins, PLCg1 and PLCg2. The PLCg isozymes have the core structure domains and a unique array of domains containing an additional PH domain, two SH2 domains and one SH3 domain. In response to extracellular stimuli, such as hormones and growth factors, receptor tyrosine kinases (RTKs) phosphorylate and activate PLCg. Activated PLCg catalyzes hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) to produce the metabolic second messenger molecules inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).Specifications
| FIGNL1 | |
| Polyclonal | |
| Unconjugated | |
| FIGNL1 | |
| fidgetin like 1, Fidgetin like protein 1, FIGNL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289777, 60530, 63979 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6GX84, Q6PIW4, Q8BPY9 | |
| Fignl1 | |
| FIGNL1 Fusion Protein Ag11709 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction