Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIH-1/HIF-1AN Antibody (12F11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FIH-1/HIF-1AN |
---|---|
Clone | 12F11 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
FIH-1/HIF-1AN Monoclonal antibody specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
FIH-1/HIF-1AN | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase | |
A synthesized peptide derived from Human FIH-1/HIF-1AN [UniProt Q9NWT6] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
12F11 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
55662 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title