Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIH-1/HIF-1AN Antibody (162c), Novus Biologicals™
SDP

Catalog No. p-4984508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130333SS 0.025 mL
NBP130333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP130333SS Supplier Novus Biologicals Supplier No. NBP130333SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

FIH-1/HIF-1AN Monoclonal specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FIH-1/HIF-1AN
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 162c
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:400
Gene Accession No. Q9NWT6
Gene Alias DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase
Gene Symbols HIF1AN
Host Species Mouse
Immunogen Full length human Factor Inhibiting HIF-1 protein [Swiss-Prot# Q9NWT6]
Molecular Weight of Antigen 40 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55662
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.