Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIH-1/HIF-1AN Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 25 publications

$206.50 - $499.50

Specifications

Antigen FIH-1/HIF-1AN
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Electron Microscopy, Knockout Validated reported in scientific literature (PMID 31937588), KnockDown Validated
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100428SS
SDP
View Documents
Novus Biologicals
NB100428SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB100428
SDP
View Documents
Novus Biologicals
NB100428
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

FIH-1/HIF-1AN Polyclonal specifically detects FIH-1/HIF-1AN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Knockout Validated.
Specifications

Specifications

FIH-1/HIF-1AN
Western Blot 1:1000-1:5000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Electron Microscopy, Knockout Validated reported in scientific literature (PMID 31937588), KnockDown Validated
Polyclonal
Rabbit
Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase
HIF1AN
IgG
Affinity Purified
Human, rat and mouse (PMID 19720742).
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Unconjugated
RUO
Human, Mouse, Rat
Q9NWT6
55662
A synthetic peptide made to the C-terminal region of human Factor Inhibiting HIF-1 (between residues 300 and the C-terminus). [UniProt# Q9NWT6]
Primary
Store at 4C. Do not freeze.
40.3 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.