Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filaggrin Antibody (FLG/1562), Novus Biologicals™

Mouse Monoclonal Antibody
$214.28 - $493.10
Specifications
| Antigen | Filaggrin |
|---|---|
| Clone | FLG/1562 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25324520
![]() |
Novus Biologicals
NBP25324520UG |
20 μg |
Each for $214.28
|
|
|||||
NB253245100
![]() |
Novus Biologicals
NBP253245100UG |
100 μg |
Each for $493.10
|
|
|||||
NB253245200
![]() |
Novus Biologicals
NBP253245200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Filaggrin Monoclonal specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Filaggrin | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 2312 | |
| Recombinant human Filaggrin protein fragment (aa 198-288) (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| FLG/1562 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
| FLG | |
| IgG1 κ | |
| Protein A or G purified | |
| Filaggrin is an intermediate filament-associated protein that aggregates keratin intermediate filaments in mammalian epidermis. It is initially synthesized as a polyprotein precursor, profilaggrin (consisting of multiple filaggrin units of 324 aa each), which is localized in keratohyalin granules, and is subsequently proteolytically processed into individual functional filaggrin molecules. Active filaggrin is present at a level of the epidermis where keratinocytes are in transition between the live nucleated granular layer and the anucleate cornified layer, suggesting that filaggrin aids in the terminal differentiation process by facilitating apoptotic machinery. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title