Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filaggrin Antibody (rFLG/1561), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308382V
Description
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Filaggrin | |
Monoclonal | |
DyLight 405 | |
ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
Recombinant human Filaggrin protein fragment (aa 198-288) (exact sequence is proprietary) (Uniprot: P20930) | |
100 μg | |
Cell Biology, Cellular Markers | |
2312 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rFLG/1561 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction