Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filaggrin Monoclonal Antibody (FLG01), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA513440
Description
MA5-13440 targets Filaggrin in IHC (P) applications and shows reactivity with Human samples. The MA5-13440 immunogen is recombinant filaggrin protein.
Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications
Filaggrin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P20930 | |
FLG | |
Recombinant filaggrin protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
FLG01 | |
Unconjugated | |
FLG | |
ATOD2; AW107830; epidermal filaggrin; filaggrin; Filaggrin (profilaggrin); FLG; ft; Pflg | |
Mouse | |
Protein G | |
RUO | |
2312 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction