Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filaggrin Mouse anti-Human, Clone: FLG/1945, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30737620UG
Description
Filaggrin Monoclonal specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Filaggrin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
ATOD2, epidermal filaggrin, filaggrin, Profilaggrin | |
Recombinant human Filaggrin protein fragment (around aa 998-1104) (exact sequence is proprietary) | |
20 μg | |
Cell Biology, Cellular Markers | |
2312 | |
Store at 4°C. | |
IgG2b κ |
Immunohistochemistry (Paraffin), Peptide Array | |
FLG/1945 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction