Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Filaggrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583129 Supplier Invitrogen™ Supplier No. PA583129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SADSSRQSGT PHAETSSGGQ AASSHEQARS SPGERHGSRH QQSADSSRHS GIPRRQASSA VRDSGHWGSS GSQASDSEGH SEESDTQSVS GHGQDGPHQQ SHQESAR Highest antigen sequence indentity to the following orthologs: Rat - 36%, Mouse - 38%.

Filaggrin is expressed only in well differentiated keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filaggrin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FLG
Gene Accession No. P20930
Gene Alias ATOD2; AW107830; epidermal filaggrin; filaggrin; Filaggrin (profilaggrin); FLG; ft; Pflg
Gene Symbols FLG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLG. Recombinant protein control fragment (Product #RP-94296).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.