Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Filamin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582234 Supplier Invitrogen™ Supplier No. PA582234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSLHETSTVL VETVTKSSSS RGSSYSSIPK FSSDASKVVT RGPGLSQAFV GQKNSFTVDC SKAGTNMMMV GVHGPKTPCE EVYVKHMGNR VYNVTYTVKE KGDY Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes one of three related filamin genes, specifically gamma filamin. These filamin proteins crosslink actin filaments into orthogonal networks in cortical cytoplasm and participate in the anchoring of membrane proteins for the actin cytoskeleton. Three functional domains exist in filamin: an N-terminal filamentous actin-binding domain, a C-terminal self-association domain, and a membrane glycoprotein-binding domain. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filamin 2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FLNC
Gene Accession No. Q14315
Gene Alias 1110055E19Rik; ABP-280; ABP280A; ABP-280-like protein; ABPA; ABPL; ABP-L; ABP-L, gamma filamin; actin binding protein 280; actin-binding protein 280; actin-binding-like protein; filamin C; Filamin C gamma; filamin C, gamma; filamin C, gamma (actin binding protein 280); filamin, gamma; filamin-2; Filamin-C; FLJ10186; FLN2; FLNC; FLN-C; gamma filamin; gamma-filamin; MFM5; MPD4
Gene Symbols FLNC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLNC. Recombinant protein control fragment (Product #RP-88627).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.