Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Filamin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596851 Supplier Invitrogen™ Supplier No. PA596851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, U-251MG, Mouse lung, Mouse spleen; Cellular Location: Cytoplasm, cell cortex, cytoskeleton Immunogen sequence: GAPGPGPADA SKVVAKGLGL SKAYVGQKSS FTVDCSKAGN NMLLVGVHGP RTPCEEILVK HVGSRLYSVS YLLKDKGEYT LVVKWGDEHI PGSPYRVVVP.

Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filamin A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Flna
Gene Accession No. P21333, Q8BTM8
Gene Alias 450 kDa protein; ABP-280; ABPX; actin binding protein 280; actin-binding protein 280; alpha filamin; alpha-filamin; CSBS; CVD1; dilated pupils 2; Dilp2; Endothelial actin-binding protein; Endothelial actin-binding protein 280); F730004A14Rik; filamin; filamin 1; filamin A; filamin A, alpha; filamin, alpha; Filamin1; filamin-1; FilaminA; Filamin-A; FLN; FLN1; Flna; FLN-A; FMD; GENA 379; gizzard filamin; MNS; NHBP; Non-muscle filamin; OPD; OPD1; OPD2; RGD1560614; XLVD; XMVD
Gene Symbols Flna
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2548-2647 of human Filamin A (NP_0011040261).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192176, 2316, 293860
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.