Learn More
Filamin A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | Filamin A |
|---|---|
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-9343
|
Bethyl Laboratories, Inc
A301133AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Promotes orthogonal branching of actin filaments and links actin filaments to membrane glycoproteins. Anchors various transmembrane proteins to the actin cytoskeleton and serves as a scaffold for a wide range of cytoplasmic signaling proteins. Interaction with FLNA may allow neuroblast migration from the ventricular zone into the cortical plate. Tethers cell surface-localized furin, modulates its rate of internalization and directs its intracellular trafficking (By similarity). Involved in ciliogenesis.Specifications
| Filamin A | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| FLN-A, actin binding protein 280, alpha-filamin, endothelial actin-binding protein, filamin 1, filamin-1, filamin-A, non-muscle filamin | |
| FLNA | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P21333 | |
| 2316 | |
| Between 400 and 450 | |
| Primary | |
| 4° C | |
| FLNA |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.