Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filamin Antibody (FLMN01 (PM6/317)), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001218
Description
Filamin Monoclonal specifically detects Filamin in Human, Mouse, Rat, Chicken, Guinea Pig, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Filamin | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1:100 - 1:500, ELISA 1:100 - 1:2000, Immunohistochemistry 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
ABP-280, ABP280A, ABP-280-like protein, ABPA, ABP-L, ABPLABP-L, gamma filamin, Actin-binding-like protein, filamin 2, filamin C, gamma, filamin C, gamma (actin binding protein 280), Filamin-2, filamin-C, FLJ10186, FLN2actin binding protein 280, FLNc, FLN-C, Gamma-filamin | |
Mouse | |
Protein G purified | |
RUO | |
2318 | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FLMN01 (PM6/317) | |
Unconjugated | |
P21333 | |
FLNC | |
Platelet filamin | |
500 μL | |
Primary | |
Human, Mouse, Rat, Chicken, Guinea Pig, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction