Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Filamin B Monoclonal Antibody (GT387)
GREENER_CHOICE

Catalog No. PIMA527748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527748 Supplier Invitrogen™ Supplier No. MA527748
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: A431, HeLa, HepG2 Predicted Reactivity: Mouse (98%), Chicken (97%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FOXK1 is expressed in these cells and regulates cell cycle progression through an interaction with its downstream target the cyclin-dependent kinase inhibitor p21 (CIP). Loss of FOXK1 in mice results in growth retardation and a severe impairment in skeletal muscle regeneration following injury. FOXK1 also shows expression in immature tissues of brain, eye, heart, lung and thymus. It also is predominantly expressed in many malignant tissues, such as tumors of the brain, colon and lymph node.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filamin B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT387
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FLNB
Gene Accession No. O75369
Gene Alias ABP-278; ABP-280; ABP-280 homolog; ABP-280-like protein; actin binding protein 278; actin-binding-like protein; AL024016; AOI; beta-filamin; Fh1; filamin B; filamin B, beta; filamin homolog 1; filamin, beta; filamin-3; FilaminB; Filamin-B; FLN1L; FLN3; FLNB; FLN-B; Larsen syndrome 1 (autosomal dominant); LRS1; SCT; TABP; TAP; thyroid autoantigen; Truncated ABP; Truncated actin-binding protein
Gene Symbols FLNB
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Filamin B. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2317, 306204
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.