Learn More
Invitrogen™ Filamin B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596132
Description
Immunogen sequence: PDGTEAEADV IENEDGTYDI FYTAAKPGTY VIYVRFGGVD IPNSPFTVMA TDGEVTAVEE APVNACPPGF RPWVTEEAYV PVSDMNGLGF KPFDLVIPFA; Positive Samples: HeLa, NIH/3T3, A-431, Mouse liver, Mouse kidney, Mouse uterus, Rat lung; Cellular Location: Cytoplasm, Z line, cell cortex, cytoskeleton, myofibril, sarcomere.
FOXK1 is expressed in these cells and regulates cell cycle progression through an interaction with its downstream target the cyclin-dependent kinase inhibitor p21 (CIP). Loss of FOXK1 in mice results in growth retardation and a severe impairment in skeletal muscle regeneration following injury. FOXK1 also shows expression in immature tissues of brain, eye, heart, lung and thymus. It also is predominantly expressed in many malignant tissues, such as tumors of the brain, colon and lymph node.
Specifications
Filamin B | |
Polyclonal | |
Unconjugated | |
FLNB | |
ABP-278; ABP-280; ABP-280 homolog; ABP-280-like protein; actin binding protein 278; actin-binding-like protein; AL024016; AOI; beta-filamin; Fh1; filamin B; filamin B, beta; filamin homolog 1; filamin, beta; filamin-3; FilaminB; Filamin-B; FLN1L; FLN3; FLNB; FLN-B; Larsen syndrome 1 (autosomal dominant); LRS1; SCT; TABP; TAP; thyroid autoantigen; Truncated ABP; Truncated actin-binding protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
2317, 286940, 306204 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75369, Q80X90 | |
FLNB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1686-1785 of human FLNB (NP_0011577891). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.