Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filamin B Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen Filamin B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-155-9546
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301726AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

FOXK1 is expressed in these cells and regulates cell cycle progression through an interaction with its downstream target the cyclin-dependent kinase inhibitor p21 (CIP). Loss of FOXK1 in mice results in growth retardation and a severe impairment in skeletal muscle regeneration following injury. FOXK1 also shows expression in immature tissues of brain, eye, heart, lung and thymus. It also is predomitly expressed in many maligt tissues, such as tumors of the brain, colon and lymph node.
Specifications

Specifications

Filamin B
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP, ABP-280 homolog, Larsen syndrome 1 (autosomal domit), actin binding protein 278, actin-binding-like protein, beta-filamin, filamin homolog 1, filamin-3, filamin-B, thyroid autoantigen
Between 2552 and 2602
Primary
4° C
FLNB
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse
O75369, Q80X90
2317, 286940
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.