Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filensin Antibody (OTI2B2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270249FR
Description
Filensin Monoclonal specifically detects Filensin in Human samples. It is validated for Western Blot.Specifications
Filensin | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
BFSP1 | |
Recombinant protein fragment corresponding to amino acids 405-665 of human Filensin (NP_001186) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2B2 | |
Western Blot | |
Beaded filament structural protein 1, beaded filament structural protein 1, filensin, CP115Lens intermediate filament-like heavy, filensin, Lens fiber cell beaded-filament structural protein CP 115, LIFL-H115 KD | |
Mouse | |
Affinity Purified | |
Vision | |
631.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction