Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Filensin Monoclonal Antibody (FIL-7B10)
GREENER_CHOICE

Catalog No. PIMA125031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125031 Supplier Invitrogen™ Supplier No. MA125031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: bovine lens. MA1-25031 will not react with mouse, rat, or chicken. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filensin
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone FIL-7B10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 15mM sodium azide
Gene BFSP1
Gene Accession No. Q06002, Q12934
Gene Alias Beaded filament structural protein 1; beaded filament structural protein 1, filensin; BFSP1; CP115; CP94; CTRCT33; cytoskeletal protein, 115 KD; filensin; Lens fiber cell beaded-filament structural protein CP 115; lens intermediate filament-like heavy; LIFL-H; LOW QUALITY PROTEIN: filensin; RP4-531H16.2
Gene Symbols BFSP1
Host Species Mouse
Immunogen Human and bovine lens filament enriched fraction (plasma membrane-cytoskeleton complex).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101121536, 281644, 631
Target Species Human, Bovine, Sheep
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.