Learn More
Invitrogen™ Filensin Polyclonal Antibody

Description
Immunogen sequence: ESESKFESES KEVSPLTQEG APEDVPDGGQ ISKGFGKLYR KVKEKVRSPK EPETPTELYT KERHVLVTGD ANYVDPRFYV SSITAKGGVA VSVAEDSVLY DG Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 58%.
Specifications
Specifications
| Antigen | Filensin |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BFSP1 |
| Gene Accession No. | Q12934 |
| Gene Alias | Beaded filament structural protein 1; beaded filament structural protein 1, filensin; BFSP1; CP115; CP94; CTRCT33; cytoskeletal protein, 115 KD; filensin; Lens fiber cell beaded-filament structural protein CP 115; lens intermediate filament-like heavy; LIFL-H; LOW QUALITY PROTEIN: filensin; RP4-531H16.2 |
| Gene Symbols | BFSP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.