Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FILIP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112954 Supplier Invitrogen™ Supplier No. PA5112954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FILIP1L Polyclonal Antibody for IHC (P), ELISA

Acts as a regulator of the antiangiogenic activity on endothelial cells. When overexpressed in endothelial cells, leads to inhibition of cell proliferation and migration and an increase in apoptosis. Inhibits melanoma growth When expressed in tumor-associated vasculature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FILIP1L
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FILIP1L
Gene Accession No. Q4L180
Gene Alias 130 kDa GPBP-interacting protein; 4631422O05Rik; 90 kDa GPBP-interacting protein; COL4A3BPIP; DOC1; DOC-1; downregulated in ovarian cancer 1-like; filamin A interacting protein 1 like; filamin A interacting protein 1-like; filamin A-interacting protein 1-like; FILIP1L; GIP130; GIP130a; GIP130b; GIP130c; GIP90; Protein down-regulated in ovarian cancer 1; protein down-regulated in ovarian cancer 1 homolog; RGD1565927
Gene Symbols FILIP1L
Host Species Rabbit
Immunogen Recombinant Human Filamin A-interacting protein 1-like protein (1-200AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11259
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.