Learn More
BrandTech™ BRAND™ Filling Tubes for seripettor™ Bottle Top Dispensers
GSA_VA
Polypropylene construction. Supplied in bulk.
$17.00 - $35.25
Specifications
Product Type Filling Tube
Material Polypropylene
For Use With (Equipment) Seripettor™ bottletop dispenser
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
03-840-94
GSA_VA
View Documents
BrandTech™ BRAND™
704534
500 mm
Pack of 2 for $35.25
Only null left
Add to Cart
03-840-95
View Documents
BrandTech™ BRAND™
704532
250 mm
Pack of 2 for $17.00
Only null left
Add to Cart
Description
Description
  • For use with seripettor™ bottletop dispensers
  • Polypropylene construction
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Filling Tube
Polypropylene
Seripettor™ bottletop dispenser
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.