Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Respirator Filter Cover for Comfort-Air™ Series
SDP

Catalog No. 19222534 Shop All Dentec Safety Specialists Inc. Products
Click to view available options
Quantity:
350/Pk.
6/Pk.

Plastic filter cover can be used with filter holder and N95 filter to mount directly to Comfort-Air™ series half and full face masks.

  • Filter cover available in pack of 6 or bulk pack of 350
  • Use with filter holder and N95 filter for use with Comfort-Air series half and full face masks
  • Allows user to conduct a positive seal check
  • Protects N95 or R95 filters from damage
  • Extends filter life
  • Prevents the user from touching the filter
  • Rugged and durable
  • Can be easily disinfected

Specifications

Color Black
Material Plastic
Product Type Filter Cover
For Use With (Equipment) Comfort-Air™ Series Half and Full Facepiece Respirators
Quantity 6/Pk.
Length (Metric) 0.635 cm
Width (Metric) 8.89 cm
Height (Metric) 8.89 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.