Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Filter Cubes for Synergy Neo2 Hybrid Multi-Mode Microplate Reader
GSA_VAGREENER_CHOICE

Catalog No. BT1035012 Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Product Type:
Alpha (96/384-well plates) Filter Cube
Dual FP Assays Filter Cube
Dual FP Green Filter Cube
Dual PMT Alpha Top Cube
Dual PMT Single Emission Upper Cube
GFP Excitation/Emission/Luminescence Filter Cube
HTRF Terbium, LanthaScreen Terbium Filter Cube
HTRF, LANCE Red Filter Cube
HTRF-Europium Filter Cube
Luminescence Standard Filter Cube
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BT1035012 HTRF Terbium, LanthaScreen Terbium Filter Cube
BT1035001 Alpha (96/384-well plates) Filter Cube
BT1035002 Dual PMT Alpha Top Cube
BT1035003 Dual PMT Single Emission Upper Cube
BT1035004 Dual FP Assays Filter Cube
BT1035011 HTRF-Europium Filter Cube
BT1035041 HTRF, LANCE Red Filter Cube
BT1035061 Dual FP Green Filter Cube
BT1035101 GFP Excitation/Emission/Luminescence Filter Cube
BT1035114 Luminescence Standard Filter Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. BT1035012 Supplier Agilent BioTek Supplier No. 1035012
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with Synergy Neo2 Hybrid Multi-Mode Microplate Reader.

For use with Synergy Neo2 Hybrid Multi-Mode Microplate Reader.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type HTRF Terbium, LanthaScreen Terbium Filter Cube
For Use With (Equipment) Synergy Neo2 Hybrid Multi-Mode Microplate Reader
Excitation/Emission EX 340/30, DM 400
For Use With (Application) Imaging
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.