Learn More
Agilent BioTek Filter Cubes for Synergy Neo2 Hybrid Multi-Mode Microplate Reader

Shop All Agilent Technologies Inc. Products


Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Dual FP Green Filter Cube |
| For Use With (Equipment) | Synergy Neo2 Hybrid Multi-Mode Microplate Reader |
| Excitation/Emission | EX 485/20, EM 528/20, DM 510 |
| For Use With (Application) | Imaging |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.