Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ FilterCart Advanced+ Replacement Filter Cartridge
Shop All Fisherbrand Products
Click to view available options
:
FilterCart Advanced+ Replacement Filter Cartridge; For Use With: FilterCart Advanced+; Color: Blue
Description
- FilterCart Advanced+ Replacement Filter Cartridge
Specifications
Specifications
Product Type | Replacement Filter |
Color | Blue |
For Use With (Equipment) | FilterCart Advanced+ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction