Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers

Catalog No. 50950254
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Emission Filter
Excitation Filter
Excitation and Emission Filter
Wavelength:
320/40 nm
335/13 nm
370/12 nm
517/15 nm
540/10 nm
590/14 nm
603/7 nm
670/10 nm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Wavelength
50-950-254 Emission Filter 590/14 nm
50-950-687 Excitation Filter 320/40 nm
50-950-688 Excitation Filter 335/13 nm
50-950-439 Excitation and Emission Filter 370/12 nm
50-950-229 Excitation and Emission Filter 517/15 nm
50-950-447 Excitation Filter 540/10 nm
50-950-258 Excitation Filter 603/7 nm
50-950-272 Excitation and Emission Filter 670/10 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 50-950-254 Supplier Thermo Scientific™ Supplier No. 1425902
Add to Cart
Add to Cart

Fluoroskan™, Fluoroskan™FL, and Luminoskan™ microplate fluorometers and luminometers can be equipped with a wide range of accessories, additional excitation filters, and emission filters to expand the instrument's functionality and application flexibility.

For running fluorometric or luminometric assays, the Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers offer reliability and optical technology that has already been proven in thousands of laboratories all over the world. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility. Additionally, the Thermo Scientific™ SkanIt™ Software increases efficiency in your lab.

  • Filters play an essential role in fluorometric measurements
  • When ordering special filters please provide as much information as possible for the best measurement results
  • If advice is needed which filters to order, contact a local Thermo Fisher Scientific representative with the following details:
    • Name of the application
    • Excitation and emission spectra
    • Peak or center wavelengths of the excitation and emission spectra
  • Fluorometric filters can also be used for luminometry depending on the application
  • For luminometric applications requiring filters, contact technical support for instructions
TRUSTED_SUSTAINABILITY

Specifications

Product Type Emission Filter
For Use With (Equipment) Fluoroskan, Fluoroskan FL
Wavelength 590/14 nm
Excitation/Emission 590 nm, HBW 14 nm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.