Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Filters for Advantage™ Cartridges

Catalog No. 19033004A Shop All MSA Products
Change view
Click to view available options
Filter Type:
N95
Connection Type:
Bayonet
Quantity:
10/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Connection Type Quantity
19-033-004A N95 Bayonet 10/Pk.
19-033-004B N95 Bayonet 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-033-004A Supplier MSA™ Supplier No. 815394
Add to Cart
Add to Cart

For use with Advantage Cartridges

Specifications

Cartridge Type Respirator Cartridge
Certifications/Compliance NIOSH
Connection Type Bayonet
Color White
Filter Type N95
Product Type Prefilter
Filter Efficiency 95%
For Use With (Application) Effective against particulate aerosols free of oil
For Use With (Equipment) Advantage™ series respirator
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.