Specifications
| Cartridge Type | Respirator Cartridge |
| Certifications/Compliance | NIOSH |
| Connection Type | Bayonet |
| Filter Type | N95 |
| Product Type | Prefilter |
| Filter Efficiency | 95% |
| For Use With (Application) | Effective against particulate aerosols free of oil |
| For Use With (Equipment) | Advantage™ series respirator |
| Quantity | 50/Pk. |