Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ fimH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144495
Description
fimH Polyclonal Antibody for Western Blot, ELISA
Involved in regulation of length and mediation of adhesion of type 1 fimbriae (but not necessary for the production of fimbriae). Adhesin responsible for the binding to D-mannose. It is laterally positioned at intervals in the structure of the type 1 fimbriae. In order to integrate FimH in the fimbriae FimF and FimG are needed.
Specifications
fimH | |
Polyclonal | |
Unconjugated | |
fimH | |
fimH; Type 1 fimbrin D-mannose specific adhesin | |
Rabbit | |
Protein G | |
RUO | |
948847 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P08191 | |
fimH | |
Recombinant Escherichia coli Type 1 fimbrin D-mannose specific adhesin (fimH) (22-300AA). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction