Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FIN 56
GREENER_CHOICESDP

Catalog No. 62805RD Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-805-RD 5 mg
62-802-5RD 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-805-RD Supplier Tocris Bioscience™ Supplier No. 6280/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ferroptosis activator

FIN 56 is a bifunctional inducer of ferroptosis in vitro. Activates squalene synthase (SQS) leading to coenzyme Q10 depletion. Also induces degradation of glutathione peroxidase 4 (GPX4), via acetyl-CoA carboxylase (ACC).

Specifications

Chemical Name or Material N2,N7-Dicyclohexyl-9-(hydroxyimino)-9H-fluorene-2,7-disulfonamide
CAS 1083162-61-1
Assay Percent Range >98%
Quantity 5 mg
Target Ferroptosis Activators
Molecular Formula C25H31N3O5S2
Formula Weight Observed MW: 517.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.