Learn More
Description
- Formed from limestone
- The high-pressure, low-heat alteration leads to massive crystalline rock
- Correlates well to Next Generation Science Standards Evidence Statements
- Ideal for classroom activities trying to ascertain differences between metamorphic rocks and their provenance
- Marble is mined heavily for use in building and architecture
Specifications
Specifications
| Product Type | Metamorphic Rock |
| Color | White |
| For Use With (Application) | Classroom activities to ascertain differences between metamorphic rocks and their provenance |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 1 Ea. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Marble, Fine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
