Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Finger Print Classification STEM Kit
GREENER_CHOICESCI_ED

Catalog No. S29559 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29559 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Carbon Black, which is known to the State of California to cause cancer.
Catalog No. S29559 Supplier Innovating Science™ Supplier No. IS3451
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Carbon Black, which is known to the State of California to cause cancer.

Innovating Science™ Finger Print Classification STEM Kit allows students to learn about the history of fingerprinting as a tool for identification.

  • Analyze data using the system that is used worldwide for classification
  • Analyze fingerprints for common characteristics and create a classification system using those characteristics
  • This kit includes an optional activity where students can practice lifting a fingerprint and comparing it to a crime scene print
  • Kit contains enough materials for 15 groups
  • Teacher's guide and Student Guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensics Class (for groups of 15)
Includes Teachers Manual and Student Guide
Quantity 1 Kit
Dimensions (L x W x H) 9 x 6.5 x 2.75 in.
Class Size 15 Lab Groups
Product Type STEM Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.