Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Nylon Gloves with Detachable Wrist Support

Catalog No. 19031845
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

A grain leather palm offers durability and abrasion resistance, while detachable wrist support protects from impact and repetitive strain injuries.

  • Stretch Nylon Lycra™ material offers breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm protects from impact
  • Fingerless style for tactile feedback and mobility
  • Grain leather offers abrasion protection and durability
  • Detachable leather wrist support prevents over-flexion and over extension to prevent repetitive strain injuries

Specifications

Cuff Material Grain Leather
Color Blue, Yellow
Hand Right
Material Nylon Lycra
Product Type Anti-impact Gloves
Size Large
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, and Maintenance
Glove Cuff Color Yellow
Series 703-20
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.