Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Finntip™ Biocon Pipette Tips
Click to view available options
Volume (Metric):
10 μL
1000 μL
250 μL
Description
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Features:
- Individually packed
- Three models in volume range 0.2-1000 μL
- Presterilized with gamma radiation
- Includes certificate of quality
Recommended for: PCR applications, molecular biology

Specifications
Specifications
Description | Finntip BioCon 10, Non-Filtered, Sterile, |
Volume (Metric) | 10 μL |
Filtered | Non-filtered |
Sterility | Sterile |
Model | Finntip BioCon 10 |
Certifications/Compliance | ISO 9001, ISO 14001, ISO 13485 |
For Use With (Equipment) | Thermo Scientific FinnTip BioCon 10 |
Product Line | FinnTip Biocon |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction