Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIP1L1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074588AF488
Description
FIP1L1 Polyclonal antibody specifically detects FIP1L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FIP1L1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp586K0717, EC 1.1.1.86, EC 4.2.1.33, Factor interacting with PAP, FIP1, FIP1 like 1 (S. cerevisiae), FIP1-like 1 protein, FLJ33619, hFip1, pre-mRNA 3'-end-processing factor FIP1, Rearranged in hypereosinophilia, Rhe | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human factor interacting with PAP using the numbering given in entry NP_112179.2 (GeneID 81608). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
81608 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction