Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIP1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596589 Supplier Thermo Scientific Supplier No. PA596589
Add to Cart
Add to Cart

FIP1L1 Polyclonal Antibody

FIP1L1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen FIP1L1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FIP1L1
Gene Accession No. Q5U317, Q6UN15, Q9D824
Gene Alias 1300019H17Rik; DKFZp586K0717; Factor interacting with PAP; factor interacting with PAPOLA and CPSF1; FIP1; FIP1 like 1; FIP1 like 1 (S. cerevisiae); FIP1 like 1b (S. cerevisiae); FIP1L1; FIP1L1 cleavage and polyadenylation specific factor subunit; fip1l1b; FIP1-like 1 protein; FLJ33619; hFip1; Pre-mRNA 3'-end-processing factor FIP1; pre-mRNA 3'-end-processing factor FIP1; LOW QUALITY PROTEIN: pre-mRNA 3'-end-processing factor FIP1; rearranged in hypereosinophilia; RHE; Rje; wu:fd36f11; zgc:103421
Gene Symbols FIP1L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human FIP1L1 (NP_0011284091).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289582, 66899, 81608
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.