Learn More
Invitrogen™ FIP200 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | FIP200 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Rb1cc1 |
| Gene Accession No. | Q8TDY2 |
| Gene Alias | 200 kDa FAK family kinase-interacting protein; 2900055E04Rik; 5930404L04Rik; ATG17; Cc1; coiled coil forming protein 1; coiled-coil-forming protein 1; FAK family kinase-interacting protein of 200 kDa; FIP200; KIAA0203; LaXp180; LOW QUALITY PROTEIN: RB1-inducible coiled-coil protein 1; phosphatase 1, regulatory subunit 131; PPP1R131; RB1 inducible coiled-coil 1; Rb1cc1; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1; RBICC |
| Gene Symbols | Rb1cc1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.