Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIP200 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528563 Supplier Invitrogen™ Supplier No. PA528563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, H1299, mouse muscle. Predicted reactivity: Human (99%), Mouse (90%), Rat (90%), Rhesus Monkey (98%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIP200
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Rb1cc1
Gene Accession No. Q8TDY2, Q9ESK9
Gene Alias 200 kDa FAK family kinase-interacting protein; 2900055E04Rik; 5930404L04Rik; ATG17; Cc1; coiled coil forming protein 1; coiled-coil-forming protein 1; FAK family kinase-interacting protein of 200 kDa; FIP200; KIAA0203; LaXp180; LOW QUALITY PROTEIN: RB1-inducible coiled-coil protein 1; phosphatase 1, regulatory subunit 131; PPP1R131; RB1 inducible coiled-coil 1; Rb1cc1; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1; RBICC
Gene Symbols Rb1cc1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human FIP200. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12421, 9821
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.