Learn More
FIP200/RB1CC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301574A
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
| FIP200/RB1CC1 | |
| Polyclonal | |
| Unconjugated | |
| RB1CC1 | |
| CC1, FIP200, 200 kDa FAK family kinase-interacting protein, FAK family kinase-interacting protein of 200 kDa, RB1-inducible coiled-coil protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9821 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8TDY2 | |
| RB1CC1 | |
| Between 250 and 300 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.