Learn More
Fire Syringe
SDPSCI_ED
Catalog No. s90538
Change view
Click to view available options
:
Boyle's Law Fire Syringe
Catalog No.
S90538 Boyle's Law Fire Syringe
1 options
Catalog No. S90538
Supplier: Science First 6121310
Includes
Aluminum piston, Lexan™ piston chamber and two custom-made O-rings
Requires
Tiny pieces of nonmoisturized tissue paper (not included).
A classic example of the Ideal Gas Law (PV=nRT).
  • When the air in the fire syringe is compressed, it can reach a temperature of over 260°C (500°F)
  • As paper burns at 235°C (454°F), the small pieces of tissue paper ignite
  • Syringe is pre-lubricated
  • After each use, blow out the carbon dust
Specifications
For Use With (Application) Demonstration of Boyle's law a classic example of the ideal Gas law (PV=nRT)
Includes Aluminum piston, Lexan™ piston chamber and two custom made O-rings
Product Type Fire Syringe