Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Modesty Curtains

Catalog No. p-35257
Click to view available options
Mounting:
Pipe
Wall

Provide privacy during emergencies

  • For use with installed safety shower system
  • Two styles available, both designed to work with any commercially available safety shower
  • Pipe-mount for freestanding, plumbed-in safety showers; measure 52 in. dia.
  • Wall-mount for freestanding systems near a wall or wall-mounted plumbed system
  • Made of 20 oz. polyester fabric with nickel-cadmium plated rings
  • Epoxy-sealed steel supports exhibit exceptional resistance to harsh environments

Specifications

Color Yellow
For Use With (Equipment) Installed Safety Shower System
Material Polyester, Nickel Cadmium, Epoxy Sealed Steel
Mounting Wall
Product Type Modesty Curtain
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.