Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisetin 96.0+%, TCI America™
SDP

Catalog No. p-7136094
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 528-48-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
MDL Number MFCD00006829
InChI Key XHEFDIBZLJXQHF-UHFFFAOYSA-N
Synonym fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
PubChem CID 5281614
ChEBI CHEBI:42567
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O

Specifications

Melting Point 330°C
Quantity 100 mg
Formula Weight 286.24
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fisetin
RTECSNumber LK9250000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.