Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ FISH Tag™ RNA Kits with Alexa Fluor™ Dyes

Catalog No. F32952
Encompass_Preferred
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647
Alexa Fluor 594
Color:
Green
Green, Red, Orange, Deep-Red
Red

FISH Tag RNA kits provide a complete solution for fluorescence in situ hybridization (FISH) applications. Choose from kits using Alexa Fluor 488, Alexa Fluor 594, or our multicolor dye kit for probe synthesis, purification, and labeling of specimens for imaging.

Choose from two, single-color FISH Tag RNA kits with Alexa Fluor dye options (green or red). The FISH RNA multicolor kit contains four spectrally distinct dyes (green, orange, red, and deep-red) to allow simultaneous localization of multiple sequence-specific RNA and DNA targets such as mRNAs in cells, tissues, and whole embryo mounts. All kits include required reagents and come with a comprehensive, detailed protocol to help ensure your success.

Complete workflow solution for FISH

FISH Tag kits employ a two-step labeling technology. First, in vitro transcription is used to enzymatically incorporate an amine-modified nucleotide into the probe template. The modified nucleotide is UTP with an NH2 group attached through a linker to the C5 position of the base. Next, dye labeling of the purified amine-modified RNA is achieved by incubation with amine-reactive dyes (e.g., Alexa Fluor dyes). These active ester compounds react with the primary amines incorporated into the probe template, covalently conjugating the dye to the modified nucleotide base. The purified probe is then ready for hybridization to the specimen. Once the hybridization assay is complete, the specimen is viewed under a fluorescence microscope to visualize the hybridized fluorescent probe.

The dyes available in the FISH Tag RNA kits are compatible with standard filter sets found on most fluorescence microscopes. Alexa Fluor dyes are brighter and more photostable than traditional fluorescent labels, providing higher resolution and improved signal-to-noise ratios compared to conventional dyes. PureLink nucleic acid purification technology is used for fast and efficient purification of the labeled probe while SlowFade Gold Antifade Mountant preserves the fluorescent signal during imaging.

See the user manual for information regarding how to calculate base:dye ratio and nucleic acid concentration, how to construct plasmids using the FISH Tag RNA kits, and about optional RNA probe fragmentation for RNA probes greater than 500 bases.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Color Green
Detection Method Fluorescence
Format Kit
Label or Dye Alexa Fluor 488
Final Product Type Probes (Labeled RNA), Probes (Labeled DNA), Oligos (Labeled)
Sample Type RNA, Tissue (FFPE), Cell Culture Cells, mRNA
Labeling Target RNA (General), siRNA
Excitation/Emission 492/520 nm
For Use With (Application) FISH Tag, fluorescent microscopy, RNA In situ hybridization
No. of Reactions 10 Reactions
Product Line Alexa Fluor™, FISH Tag™
Product Type RNA In Situ Hybridization Assay Kit
Quantity 1 kit
Target RNA
Target Specificity Not Target-Specific
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.