Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ exACTGene™ DNA Ladders

Catalog No. bp2576100 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Application):
General purpose, large digested DNA
General purpose, wide size range
Large size PCR or cloning
No. of Bands:
10
13
19
Product Type:
DNA Molecular Weight Marker/Ladder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) No. of Bands Product Type
BP2576100 Large size PCR or cloning 10 DNA Molecular Weight Marker/Ladder
BP2578100 General purpose, large digested DNA 13 DNA Molecular Weight Marker/Ladder
BP2579-100 General purpose, wide size range 19 DNA Molecular Weight Marker/Ladder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2576100

Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

  • Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
  • Ideal for size assessment and quantitative determination of DNA mass
  • Contain loading dye to reduce pipetting steps and save time
  • Have two or three high intensity reference bands for better sizing
  • Provided in sufficient quantity to load 100 lanes

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Form Liquid
Formulation Premixed with loading dye
For Use With (Application) Large size PCR or cloning
No. of Bands 10
Quantity 100 Lanes
Storage Requirements Stable at room temperature for two years.
Sufficient For 100 Lanes
Product Type DNA Molecular Weight Marker/Ladder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.